Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Low Retention Pipette Tips, Racked

Catalog No. 21402679
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
300 μL
1000 μL
1250 μL
Sterility:
Non-sterile
Sterile
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
21-402-679 10 μL Non-sterile
21-402-685 200 μL Non-sterile
21-402-688 200 μL Sterile
21-402-689 300 μL Non-sterile
21-402-690 1000 μL Non-sterile
21-402-692 1000 μL Sterile
21-402-693 1250 μL Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 21-402-679 Supplier Bio Plas™ Supplier No. 3100
Add to Cart
Add to Cart

Racked, Sterile, Natural Color, Polypropylene, Made In The USA

  • Low retention design reduces sample loss and enhances accuracy for precise liquid handling
  • Pre-racked format supports organized storage and convenient access during laboratory workflows
  • Sterile tips help maintain contamination-free conditions for sensitive applications
  • Natural color improves visibility of liquids for better pipetting control
  • Manufactured from high-quality polypropylene for durability and chemical resistance, made in the USA

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Flat
Quantity 96 Tips/Rack
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.