Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ LTS Style Pipette Tip

Catalog No. 21402656
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Rack
Refill
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
21-402-656 Non-sterile Rack
21-402-657 Sterile Rack
21-402-655 Non-sterile Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-402-656 Supplier Bio Plas™ Supplier No. 2200R
Add to Cart
Add to Cart

Designed for secure sealing, easy attachment, and smooth ejection with compatible pipettes

  • Designed in LTS style to ensure a secure seal and easy attachment and ejection with compatible pipettes
  • Supports precise and consistent liquid handling across various laboratory applications
  • Engineered for smooth performance to reduce user effort during repetitive pipetting tasks
  • Compatible with LTS-style pipette systems for reliable functionality
  • Suitable for routine laboratory use requiring accuracy and efficiency

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Flat
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.