Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ LTS Style Pipette Tip

Catalog No. 21402660
Change view
Click to view available options
Volume (Metric):
200 μL
300 μL
Sterility:
Non-sterile
Sterile
Format:
Rack
Refill
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Format
21-402-660 200 μL Non-sterile Rack
21-402-659 200 μL Non-sterile Refill
21-402-661 200 μL Sterile Rack
21-402-668 300 μL Non-sterile Rack
21-402-667 300 μL Non-sterile Refill
21-402-669 300 μL Sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21-402-660 Supplier Bio Plas™ Supplier No. 2202R
Add to Cart
Add to Cart

Racked for organized storage and easy access during laboratory use

  • LTS style design ensures a secure seal and effortless attachment and ejection with compatible pipettes
  • Racked format allows for convenient handling, organization, and efficient workflow in the lab
  • Supports accurate and consistent liquid handling for a variety of applications
  • Engineered for smooth operation to reduce hand strain during repetitive pipetting tasks
  • Compatible with LTS-style pipette systems for dependable performance

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Flat
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.