Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Standard Pipette Tips, Bulk, Beveled

Catalog No. 21402675
Change view
Click to view available options
Color Code:
White
Yellow
Format:
Bagged
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code Format
21-402-675 Yellow Bagged
21-402-673 White Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-675 Supplier Bio Plas™ Supplier No. 3003
Add to Cart
Add to Cart

Universal fit pipette tips with a beveled design, supplied in bulk packaging

  • Universal fit design ensures compatibility with a wide range of pipette brands and models
  • Beveled tip enhances accuracy and ease of dispensing, especially for precise liquid handling
  • Bulk packaging provides a cost-effective solution for high-volume laboratory use and storage efficiency
  • Engineered for consistent performance across routine laboratory applications
  • Ideal for general-purpose pipetting tasks requiring reliability and versatility

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Bagged
Tip Style Beveled
Product Type Pipette Tip
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.