Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Standard Pipette Tips, Gilson Fit

Catalog No. 21402671
Change view
Click to view available options
Volume (Metric):
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-402-671 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-402-671 Supplier Bio Plas™ Supplier No. 3000
Add to Cart
Add to Cart

Designed for Gilson compatibility, supplied in a convenient bulk pack for efficient laboratory use.

  • Designed for accurate 10 μL pipetting with reliable Gilson compatibility
  • Bulk pack format supports high-volume laboratory workflows and cost efficiency
  • Ensures consistent performance for precise liquid handling tasks
  • Suitable for routine laboratory applications requiring small-volume transfers

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Bagged
Tip Style Flat
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.