Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Standard Pipette Tips, Racked, Beveled

Catalog No. 21402676
Change view
Click to view available options
Color Code:
White
Yellow
Format:
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code Format
21-402-676 Yellow Rack
21-402-674 White Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-676 Supplier Bio Plas™ Supplier No. 3003R
Add to Cart
Add to Cart

Beveled pipette tips in a racked format, supplied with 96 tips per rack for convenient laboratory use

  • Beveled tip design improves precision and control during liquid dispensing tasks
  • Racked packaging provides organized storage and easy access, enhancing workflow efficiency
  • Includes 96 tips per rack, suitable for routine laboratory applications
  • Compatible with a wide range of pipettes for versatile use
  • Designed for consistent performance and reliable results in everyday pipetting

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Beveled
Quantity 96 Tips/Rack
Product Type Pipette Tip
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.