Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Standard Pipette Tips

Catalog No. 21402678
Change view
Click to view available options
Format:
Bagged
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
21-402-678 Rack
21-402-677 Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-678 Supplier Bio Plas™ Supplier No. 3004R
Add to Cart
Add to Cart

Designed for compatibility with a wide range of pipettes

  • 1000 μL volume capacity suitable for larger liquid handling applications in the laboratory
  • Universal fit design ensures compatibility with most major pipette brands and models
  • Engineered for accurate and consistent liquid transfer across routine workflows
  • Supports reliable performance in general-purpose laboratory pipetting tasks
  • Designed for ease of use and dependable handling in everyday applications

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear Blue
Autoclavable Autoclavable
Format Rack
Tip Style Flat
Quantity 96 Tips/Rack
Product Type Pipette Tip
Color Code Blue
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.