Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Wide Bore Pipette Tip

Catalog No. 21402719
Change view
Click to view available options
Volume (Metric):
200 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-402-719 1000 μL
21-402-717 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-719 Supplier Bio Plas™ Supplier No. 3401R
Add to Cart
Add to Cart

Racked and graduated, supplied with 96 tips per rack for convenient laboratory use

  • Wide bore design enables gentle handling of viscous or sensitive samples, minimizing shear stress
  • Graduated markings provide improved volume estimation and greater pipetting accuracy
  • Racked format ensures organized storage and easy accessibility during workflows
  • Supplied with 96 tips per rack, supporting efficient routine laboratory operations
  • Designed for consistent and reliable performance in various liquid handling applications

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Flat
Quantity 96 Tips/Rack
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.