Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ XL Low Retention Pipette Tips, EnviroPak Refills

Catalog No. 21402684
Change view
Click to view available options
Volume (Metric):
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-402-684 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-402-684 Supplier Bio Plas™ Supplier No. 3101E
Add to Cart
Add to Cart

Extra long and supplied as EnviroPak refills, racked with 96 tips per rack, made from natural polypropylene in the USA.

  • Designed for precise 10 μL pipetting with XL extra-long length for extended reach applications
  • Low retention surface minimizes sample loss and improves accuracy
  • EnviroPak refill system supports reduced waste and efficient lab operations
  • Supplied racked with 96 tips per rack for organized and convenient use
  • Manufactured from high-quality natural polypropylene for durability and performance
  • Natural color enhances visibility during pipetting tasks
  • Made in the USA ensuring reliable quality and consistency

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Refill
Tip Style Flat
Quantity 96 Tips/Rack
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.