Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ XL Low Retention Pipette Tips

Catalog No. 21402683
Change view
Click to view available options
Format:
Bulk
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
21-402-683 Bulk
21-402-682 Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-683 Supplier Bio Plas™ Supplier No. 3101B
Add to Cart
Add to Cart

Extra long, low retention pipette tips in natural color, made from polypropylene and manufactured in the USA

  • Extra long design allows access to deep or narrow vessels, improving reach and usability
  • Low retention surface reduces sample loss and enhances accuracy, especially for small or critical volumes
  • Natural color provides clear visibility of liquids during pipetting procedures
  • Constructed from high-quality polypropylene for durability and strong chemical resistance
  • Made in the USA, ensuring consistent quality and reliable laboratory performance

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Bulk
Tip Style Flat
Quantity 1000 Tips/Bag
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.