Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ XL LTS Style Pipette Tips

Catalog No. 21402665
Change view
Click to view available options
Format:
Rack
Refill
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Sterility
21-402-665 Rack Sterile
21-402-664 Rack Non-sterile
21-402-663 Refill Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-402-665 Supplier Bio Plas™ Supplier No. 2203RS
Add to Cart
Add to Cart

Designed for extended reach and compatibility with LTS pipette systems

  • 1000 μL capacity suitable for larger volume liquid handling applications
  • XL length design enables access to deep or narrow lab vessels with ease
  • LTS style ensures a secure seal with effortless attachment and ejection
  • Compatible with LTS pipette systems for reliable and consistent performance
  • Engineered for accurate and efficient pipetting in routine laboratory workflows

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Flat
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.