Learn More
Biotium MitoView™ Green, Trial Size (50 UG)
Supplier:  Biotium 70054T
Encompass_Preferred
MitoView™ dyes are membrane-permeant fluorogenic stains that rapidly accumulate in mitochondria of live cells. They can be imaged without washing, and are available with blue, green, far-red, and near-infrared fluorescence.MitoView™ Green is a green fluorescent mitochondrial dye (Ex/Em 490/520 nm) similar to MitoTracker Green FM. The dye is non-fluorescent until it paritions into the mitochondrial membrane. The staining relies on mitochondrial mass, not on mitochondrial membrane potential (MMP) and can therefore be used in both live cells and fixed cells. Biotium also offers several classical dyes for measuring MMP including JC-1, a ratiometric dye that forms red aggregates in healthy cells and green monomers when MMP is lost; and TMRM and TMRE, red fluorescent dyes used to quantitatively measure mitochondrial membrane potential using the Nernst equation.MitoTracker is a registered trademark of Thermo Fisher Scientific.
Catalog No. NC1497579