Learn More
Assaypro Biotinylated Human Complement C1q, 100 μg, 410 kDa, lyophilized powder, >96% purity, 10 mM HEPES, 300 mM NaCl, pH 7.2, 5% Trehalose, for research applications
Catalog No. NC2680851
Encompass
Biotinylated Human Complement C1q is a highly purified protein derived from human plasma, designed for research applications in immunology and biochemistry. This 410 kDa protein is biotinylated, which enhances its utility for sensitive detection in various assays. It is particularly important for exploring the classical complement pathway and its role in autoimmune diseases such as Lupus (SLE). Supplied in a lyophilized format, it offers stability and consistent performance for diverse experimental needs.
  • High purity >96% by SDS-PAGE
  • Molecular weight of 410 kDa
  • Presented as lyophilized powder
  • Formulated with 10 mM HEPES, 300 mM NaCl, pH 7.2, and 5% trehalose
  • Stable storage at -20°C or below
  • Product size of 100 μg
  • Reactive to human species
  • Associated with autoimmune diseases like Lupus (SLE)
Catalog No. NC2680851 Supplier Assaypro Supplier No. PHC11211BIO
Biotinylated Human Complement C1q is a highly purified protein derived from human plasma, designed for research applications in immunology and biochemistry. This 410 kDa protein is biotinylated, which enhances its utility for sensitive detection in various assays. It is particularly important for exploring the classical complement pathway and its role in autoimmune diseases such as Lupus (SLE). Supplied in a lyophilized format, it offers stability and consistent performance for diverse experimental needs.
  • High purity >96% by SDS-PAGE
  • Molecular weight of 410 kDa
  • Presented as lyophilized powder
  • Formulated with 10 mM HEPES, 300 mM NaCl, pH 7.2, and 5% trehalose
  • Stable storage at -20°C or below
  • Product size of 100 μg
  • Reactive to human species
  • Associated with autoimmune diseases like Lupus (SLE)