Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems AcroBiosystems Biotinylated Mouse Integrin Alpha V Beta 3 Heterodimer Protein, 25 μg (IT3-H82W3)

Catalog No. NC3762857
Encompass_Preferred

Biotinylated mouse integrin alpha V beta 3 heterodimer protein for binding, detection, and assay development. Expressed in HEK293 cells and enzymatically biotinylated via Avitag, the protein is supplied lyophilized in a stabilized buffer and verified by SEC-MALS for molecular weight and oligomeric state.

  • Contains 25 μg of lyophilized protein.
  • Expression host HEK293 cells.
  • Tags include His, Avitag, and tag free.
  • Amino acid ranges ITGAV Phe 31-Val 988 and ITGB3 Glu 26-Asp 717.
  • Molecular weight approximately 200-240 kDa by SEC-MALS.
  • Purity greater than 95% by SDS-PAGE.
  • Endotoxin less than 1.0 EU per μg (LAL).
  • Formulation lyophilized from 50 mM Tris, 150 mM NaCl, pH 7.5, with trehalose.
  • Storage long-term at -20°C or lower in the lyophilized state.

Catalog No. NC3762857 Supplier ACROBiosystems Supplier No. IT3H82W325UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated mouse integrin alpha V beta 3 heterodimer protein for binding, detection, and assay development. Expressed in HEK293 cells and enzymatically biotinylated via Avitag, the protein is supplied lyophilized in a stabilized buffer and verified by SEC-MALS for molecular weight and oligomeric state.

  • Contains 25 μg of lyophilized protein.
  • Expression host HEK293 cells.
  • Tags include His, Avitag, and tag free.
  • Amino acid ranges ITGAV Phe 31-Val 988 and ITGB3 Glu 26-Asp 717.
  • Molecular weight approximately 200-240 kDa by SEC-MALS.
  • Purity greater than 95% by SDS-PAGE.
  • Endotoxin less than 1.0 EU per μg (LAL).
  • Formulation lyophilized from 50 mM Tris, 150 mM NaCl, pH 7.5, with trehalose.
  • Storage long-term at -20°C or lower in the lyophilized state.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.