Learn More
Cayman Chemical Biotoxis qPCR Detection Kit
SDP
Catalog No. 502939683
Encompass_Preferred
A qPCR all-in-one detection kit that detects B. anthracis, Y. pestis and F. tularensis.
Catalog No. 50-293-9683 Supplier Cayman Chemical Supplier No. 283221 EA
A qPCR all-in-one detection kit that detects B. anthracis, Y. pestis and F. tularensis.