Learn More
Sigma Aldrich Fine Chemicals Biosciences BioTracker 488 Green Lipid
Catalog No. 502906591
Encompass
Lipids are fundamental building blocks of cells and play important cellular roles. Lipids are key components of the plasma membrane and other cellular compartments including the nuclear membrane endoplasmic reticulum Golgi apparatus and trafficking vesicles such as endosomes and lysosomes. Intracellular lipid droplets are cytoplasmic organelles involved in the storage and regulation of triglycerides and cholesterol esters. Lipids have been implicated in various diseases including heart vascular and neurological diseases.BioTracker Lipid dyes rapidly stain lipid droplets in live or fixed cells with no wash step and minimal background staining of cellular membranes or other organelles. Cells also can be fixed and permeabilized after staining.The BioTracker 488 Green Lipid Dye has an excitation around 430 nm and can be excited equally well at 405 nm or 488 nm. In cells it stains lipid droplets with bright green fluorescence using the fITC detection channel.Spectral PropertiesAbsorb
Catalog No. 50-290-6591 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCT144
Lipids are fundamental building blocks of cells and play important cellular roles. Lipids are key components of the plasma membrane and other cellular compartments including the nuclear membrane endoplasmic reticulum Golgi apparatus and trafficking vesicles such as endosomes and lysosomes. Intracellular lipid droplets are cytoplasmic organelles involved in the storage and regulation of triglycerides and cholesterol esters. Lipids have been implicated in various diseases including heart vascular and neurological diseases.BioTracker Lipid dyes rapidly stain lipid droplets in live or fixed cells with no wash step and minimal background staining of cellular membranes or other organelles. Cells also can be fixed and permeabilized after staining.The BioTracker 488 Green Lipid Dye has an excitation around 430 nm and can be excited equally well at 405 nm or 488 nm. In cells it stains lipid droplets with bright green fluorescence using the fITC detection channel.Spectral PropertiesAbsorb