Learn More
Sigma Aldrich Fine Chemicals Biosciences BioTracker 488 Green Mitochondria Dye Live cell imaging mitochondrial dye that stains the membrane of mitochondria used to detect cell viability, metabolic activity and overall cell health. |
Catalog No. NC2001149
Encompass
BioTracker 488 Green Mitochondria Dye Live cell imaging mitochondrial dye that stains the membrane of mitochondria used to detect cell viability, metabolic activity and overall cell health. |
Catalog No. NC2001149 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCT136
BioTracker 488 Green Mitochondria Dye Live cell imaging mitochondrial dye that stains the membrane of mitochondria used to detect cell viability, metabolic activity and overall cell health. |