Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTracker 510 Green β-Glu Dye, MilliporeSigma™

Catalog No. SCT026 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT026 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT026 Supplier MilliporeSigma Supplier No. SCT026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye for Beta-glucosidase (β-Glu).

  • Beta-glucosidase (β-Glu) catalyzes the hydrolysis of the glycosidic bonds to terminal non-reducing residues in beta-D-glucosides and oligosaccharides, with release of glucose.
  • The BioTracker 510 Green β-Glu Dye is permeable through the cell membrane and is fluorescent substrate 9- (4′-methoxy-2′-methylphenyl) -6- (β-D-glucopylanosyloxy) -xanthen-3-one for detecting β-glucosidase in living cells. Non-fluorescent dye is hydrolyzed by the β-Glucosidase, and generates bright green fluorescence (510 nm) when it is irradiated by the 490 nm excitation light.
  • Absorbance: 492 nm
  • Emission: 510 nm
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-(4'-Methoxy-2'-methylphenyl)-6-(beta-D-glucopyranosyloxy)-3H-xanthen-3-one
Absorbance 492 nm
CAS 67-68-5
Quantity 1 mg
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.