Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTracker 515 Green ONOO- Dye, MilliporeSigma™

Catalog No. SCT035 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT035 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT035 Supplier MilliporeSigma Supplier No. SCT035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye for peroxynitrite (ONOO-) used to detect reactive oxygen species (ROS) generated from the interaction of nitric oxide (NO) and superoxide (O).

  • Peroxynitrite (ONOO-) is a strong oxidant generated from the interaction of nitric oxide (NO) and superoxide (O). The in vivo formation of this compound has recently been demonstrated in macrophages, neutrophils and endothelial cells. The role of ONOO- in inflammatory diseases is further supported by the detection of immunoreactive nitrotyrosine on proteins at sites of inflammation.
  • The BioTracker 515 Green ONOO- Dye is a fluorescent probe that detects peroxynitrite (ONOO-) in living cells. Fluorescent intensity is not increased by the existence of hydroxyl radical, singlet oxygen, hydrogen peroxide, hypochlorite, nitric oxide or superoxide.
  • Absorbance: 490 nm
  • Emission: 515 nm
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NiSPY-3(Nitrative Stress Sensing Pyrromethene Dye)
Absorbance 490 nm
Color Orange
Quantity 1 mg
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.