Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTracker 519 Green β-Gal Dye, MilliporeSigma™

Catalog No. SCT025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 x 30 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT025 5 x 30 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT025 Supplier MilliporeSigma Supplier No. SCT025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye for β-galactosidase (β-gal) used to detect LacZ reporter gene expression and cellular senescence (SA-β-gal).

  • β-galactosidase, also called beta-gal or β-gal, is a glycoside hydrolase enzyme that catalyzes the hydrolysis of β-galactosides into monosaccharides through the breaking of a glycosidic bond. β-gal is commonly used in molecular biology as a reporter marker to monitor gene expression using the chromogenic X-Gal. β-gal has also been used to measure cellular senescence (SA-β-gal).
  • The BioTracker 543 Green β-Gal Dye is a fluorescent probe for the detection of β-galactosidase in living cells. It can be applied to fluorescent imaging and selection of cell and tissue transfected with lacZ along as a measurement of cellular senescence. Other applications include gene analysis by fluorescent imaging, monitoring transfection efficiency, and the study of gene promoter or enhancer elements. The dye can stain LacZ expressing HEK293 cells both before and after 3% paraformaldehyde fixation.
  • Absorbance: 497 nm
  • Emission: 519 nm
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 497 nm
CAS Yellow
Quantity 5 x 30 nmol
Molecular Weight (g/mol) 561.50
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.