Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTracker 520 Green Hypoxia Dye, MilliporeSigma™

Catalog No. SCT033 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 x 25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT033 5 x 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT033 Supplier MilliporeSigma Supplier No. SCT033
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye for hypoxia that increases in fluorescence intensity with decreasing oxygen levels.

  • Hypoxia is defined as the reduction or lack of oxygen in organs, tissues, or cells. Hypoxia in living tissue closely relates to a wide array of pathologies, including cancer and ischemia. Existing hypoxia detection probes, such as pimonidazole, require cell fixation followed by immunostaining.
  • The BioTracker 520 Green Hypoxia Dye is a fluorescent imaging probe for the detection of hypoxia in living cells. This probe is as sensitive as pimonidazole and can be used for live cell fluorescence imaging and flow cytometry applications.
  • Absorbance: 498 nm
  • Emission: 520 nm
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 498 nm
CAS 67-68-5
Quantity 5 x 25 μg
Molecular Weight (g/mol) 582.59
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.