Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTracker 525 Green GGT Dye, MilliporeSigma™

Catalog No. SCT028 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 x 20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT028 10 x 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT028 Supplier MilliporeSigma Supplier No. SCT028
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye for Gamma-glutamyltransferase (GGT) for cancer research applications.

  • Gamma-glutamyltransferase, or GGT, is an enzyme that plays a key role in the gamma-glutamyl cycle, a pathway for the synthesis and degradation of glutathione and drug and xenobiotic detoxification. GGT is predominantly used as a diagnostic marker for liver disease and is often significantly increased in human malignancies and cancers.
  • The BioTracker 525 Green GGT Dye is cancer-selective fluorescent imaging probe. This probe is rapidly activated by g-glutamyltranspeptidase (GGT), which is known to be tumor-associated enzyme, to give highly fluorescent compound. The membrane-permeable dye is applicable to live cell or in vivo imaging of cancer cells.
  • Absorbance: 496 nm
  • Emission: 525 nm
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ProteoGREEN-gGlu
Absorbance 496 nm
Color Orange
Quantity 10 x 20 μg
Molecular Weight (g/mol) 558.48
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.