Learn More
Description
- Lysosomes are membrane-enclosed organelles that contain an array of enzymes capable of breaking down all types of biological material including proteins, nucleic acids, carbohydrates, and lipids. Lysosomes function as the digestive system of the cell, serving both to degrade material taken up from outside the cell and to digest obsolete components of the cell itself.
- The BioTracker 560 Orange Lysosome Dye is a live cell fluorescence imaging probe which enhances fluorescence dramatically in acidic environments. This probe can stain acidic organelles such as lysosome, late endosome and granule selectively. Namely, in the condition of pH 5.0, identical to the environment of acidic organelles, fluorescence intensity is 50-fold or more compared to physical pH 7.4. The BioTracker 560 Orange Lysosome Dye shows great photostability against irradiating excitation light. As it emits orange color fluorescent by excitation at 532 nm or 514 nm, multicolor imaging is possible by combining with blue fluorescence (CFP, Hoechst, etc.), green fluorescence (GFP, fluorescein, etc.) and near-infrared fluorescence.
- Absorbance: 535 nm
- Emission: 560 nm
Specifications
Specifications
| Absorbance | 535 nm |
| CAS | 1616501-21-3 |
| pH | 5 to 7.4 |
| Quantity | 10 x 10 μg |
| Molecular Weight (g/mol) | 703.47 |
| Grade | Research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.