Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTracker 625 Red GSH Dye, MilliporeSigma™

Catalog No. SCT036 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 x 25 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT036 2 x 25 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT036 Supplier MilliporeSigma Supplier No. SCT036
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye for glutathione (GSH) that detects reactive oxygen species (ROS) free radicals.

  • Glutathione (GSH) is an important antioxidant preventing damage to important cellular components caused by reactive oxygen species (ROS) including free radicals, peroxides, lipid peroxides, and heavy metals. Alterations in glutathione homeostasis are associated with a variety of diseases; therefore, real-time live-cell imaging and quantification of GSH dynamics are important for understanding pathophysiological processes.
  • The BioTracker 625 Red GSH Dye is probe for glutathione (GSH) in living cells. The dye is formed with an orange fluorescent dye (T) and a deep-red fluorescent dye (S). At a low GSH concentration, green light excitation of T, leads to an energy transfer to S and a subsequent emission of deep-red fluorescent light. At a high GSH concentration, the energy transfer does not occur, so the probe emits the orange fluorescent light from the emission of T. The dye produces a quick and reversible response to the GSH concentration changes, relatively slow photobleaching and low cytotoxicity.
  • Absorbance: 520 nm
  • Emission: 625 nm and 582 nm
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 520 nm
Quantity 2 x 25 nmol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.