Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BioTracker Cys Hcy GSH Live Cell Dye Probe 1

Catalog No. SCT055MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT055MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT055MI Supplier MilliporeSigma™ Supplier No. SCT055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye to simultaneously report cellular levels of GSH, Cys, Hcy

  • Live cell imaging dye to simultaneously report for cellular levels of Glutathione (GSH), Cysteine (Cys) and Homocysteine (Hcy) which are inter-related and implicated in several diseases
  • Uses:
    • Used for simultaneous detection of GSH and Cys/Hcy in living cells and tissues
    • GSH is detected in the red channel while Cys/Hcy are detected in the blue channel
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity 1 mg
    Content And Storage Store -20°C, dessicate and protect from light
    Detection Method Fluorescence
    Quantity 1 mg
    Solution Type Stain
    Emission GSH 587 nm, Cys/Hcy 454 nm
    For Use With (Application) Cell Based Assays
    Form Solid

    For research purposes only

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.