Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BioTracker Cys Hcy GSH Triple Live Cell Dye

Catalog No. SCT062MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT062MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT062MI Supplier MilliporeSigma™ Supplier No. SCT062
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye to simultaneously report cellular levels GSH, Cys, Hcy

  • Uses:
    • Live cell imaging dye to simultaneously report for cellular levels of Glutathione (GSH), Cysteine (Cys) and Homocysteine (Hcy) which are inter-related and implicated in several diseases
    • A novel fluorescent probe with four binding sites, which can simultaneously and selectively detect Cys, Hcy, and GSH in three different emission channels
    • Used simultaneously monitor endogenous Cys and GSH and exogeneous Cys, Hcy, and GSH through multicolor imaging
TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 mg
Content And Storage Store -20°C, dessicate and protect from light
Quantity 1 mg
Solution Type Stain
Emission 457 nm (Blue, Cys), 559 nm (Red, Hcy), 529 nm (Green, GSH)
Excitation 360 nm (Cys), 480 nm (Hcy), 400 nm (GSH)
For Use With (Application) Cell based assays
Form Powder

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.