Learn More
Description
- Selective for GSH in the red region and for cysteine/homocysteine (Cys/Hcy) in the green region
- When treated with GSH, DACP-2 showed strong fluorescence enhancement in comparison to that for closely related species such as amino acids, including Cys/Hcy
Specifications
Specifications
| Content And Storage | Store -20°C, dessicate and protect from light |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| Solution Type | Stain |
| Emission | Emission 590 nm for GSH, 510 nm for Cys |
| For Use With (Application) | Cell Based Assays |
| Form | Solid |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.