Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BioTracker ERthermAC Temperature-Sensitive Live Cell Dye

Catalog No. SCT057MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT057MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT057MI Supplier MilliporeSigma™ Supplier No. SCT057
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging thermosensitive dye to visualize heat production in brown adipocytes

  • Small size, excellent cellular uptake, low cytotoxicity and robust photostability make it very biocompatible
  • Data obtained using this dye comparable to data obtained by conventional methods measuring mitochondrial depolarization, oxygen consumption, and extracellular acidification rates, and with thermal power measurements obtained using calScreener
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store -20°C, dessicate and protect from light
Detection Method Fluorescence
Quantity 1 mg
Solution Type Stain
Emission 590 (Orange/Red) in polarized mitochondria, 527 nm (Green) in depolarized mitochondria
For Use With (Application) Cell Based Assays
Form Solid

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.