Learn More
BioTracker Far-Red Peroxynitrite Live Cell Dye, MilliporeSigma™
Catalog No. SCT052MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
SCT052MI 1 mg
1 options
Catalog No. SCT052MI Supplier MilliporeSigma Supplier No. SCT052
Add to Cart
Add to Cart
Live cell imaging probe for detection and imaging peroxynitrite
  • Can be used for ONOO- detection
  • Probe is cell permeable and displays a high sensitivity and selectivity towards peroxynitrite over other ROS/RNS
  • Can be used to detect exogenous and endogenous peroxynitrite in living systems and displays long wavelength absorption (600 nm) and emission (638 nm) bands
TRUSTED_SUSTAINABILITY
Specifications
Absorption Ratio 600 nm
Molecular Weight (g/mol) 480.6
Quantity 1 mg
For research purposes only