Learn More
Description
- Uses:
- To study neurodegenrative diseases such as Alzheimer's disease
- BioTracker IRI-1 b-Amyloid plaque live cell dye is a near-infrared-emissive two-photon microscopy (TPM) dye with high affinity for Aβ-fibrils
Specifications
Specifications
| Capacity | 1 mg |
| Content And Storage | Store -20°C, dessicate and protect from light |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| Solution Type | Stain |
| Emission | 580 to 779 nm |
| Excitation | 850 |
| For Use With (Application) | Cell Based Assays, Fluorescence, Neuroscience |
| Form | Powder |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.