Learn More
Sigma Aldrich Fine Chemicals Biosciences BioTracker NIR694 Nuclear
Catalog No. 502906518
Encompass
The nucleus is a membrane-enclosed organelle found in eukaryotic cells. Cell nuclei contain most of the cells genetic material organized as multiple long linear DNA molecules in complex with a large variety of proteins such as histones to form chromosomes. Traditionally DNA stains such as DAPI and Hoechst have been used for microscopy but require fixation and cannot be used for live cell imaging.The BioTracker NIR694 Nuclear Dye (Water) is a far red nuclear stain for live cells. The dye can be excited by wavelengths from 488 to 647nm and emits far red fluorescence with emission maximum at 694nm. The dye specifically stains the nucleus of live cells. Staining does not require a wash step and demonstrates greater photostability than the traditional blue fluorescent nuclear stains DAPI and Hoechst. also can be used to stain live cells for cell cycle distribution analysis by flow cytometry BioTracker NIR694 Nuclear Dye (Water) staining is not fixable. for fixed cells we recommend the B
Catalog No. 50-290-6518 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCT117
The nucleus is a membrane-enclosed organelle found in eukaryotic cells. Cell nuclei contain most of the cells genetic material organized as multiple long linear DNA molecules in complex with a large variety of proteins such as histones to form chromosomes. Traditionally DNA stains such as DAPI and Hoechst have been used for microscopy but require fixation and cannot be used for live cell imaging.The BioTracker NIR694 Nuclear Dye (Water) is a far red nuclear stain for live cells. The dye can be excited by wavelengths from 488 to 647nm and emits far red fluorescence with emission maximum at 694nm. The dye specifically stains the nucleus of live cells. Staining does not require a wash step and demonstrates greater photostability than the traditional blue fluorescent nuclear stains DAPI and Hoechst. also can be used to stain live cells for cell cycle distribution analysis by flow cytometry BioTracker NIR694 Nuclear Dye (Water) staining is not fixable. for fixed cells we recommend the B