Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BioTracker Tau filaments pFTAA Live Cell Dye

Catalog No. SCT066MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT066MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT066MI Supplier MilliporeSigma™ Supplier No. SCT066
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live cell imaging dye for detection of disease-association protein aggregates

  • A novel fluorescent probe that can bind to and discriminate between amyloid-b (Aβ) plaques and tau neurofibrillary tangles (NFTs) in Alzheimer's disease (AD)-like pathology in transgenic mouse brain tissue sections
TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 mg
Content And Storage Store -20°C, dessicate and protect from light
Detection Method Fluorescence
Quantity 1 mg
Solution Type Stain
Emission 550 nm
Excitation 405 nm
For Use With (Application) Cell Based Assays
Form Powder

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.