Learn More
AirSea USA Biotransporter 2 - 1.5L UN Combination packaging, 4H2/CLASS 6.2
Catalog No. 501534827
Encompass_Preferred
Category A infectious packaging. Biotransporter 2 combination packaging is a white plastic outer case with carry handle containing an insulating polystyrene box. It is supplied as complete combination packagin, including: 1 x 1.5L Biojar (code 490), absorbent cushioning material and carry handle. An empty box can contain up to 7kg of dry ice and can give up to 130 hours protection. Loss rates were conducted a room temperature and are provided as guidelines only. This product is delivered fully assembled with instuctions and hazard labels.
Catalog No. 50-153-4827 Supplier AirSea USA Supplier No. 570CASE
Category A infectious packaging. Biotransporter 2 combination packaging is a white plastic outer case with carry handle containing an insulating polystyrene box. It is supplied as complete combination packagin, including: 1 x 1.5L Biojar (code 490), absorbent cushioning material and carry handle. An empty box can contain up to 7kg of dry ice and can give up to 130 hours protection. Loss rates were conducted a room temperature and are provided as guidelines only. This product is delivered fully assembled with instuctions and hazard labels.