Learn More
AirSea USA Biotransporter Cargo - 12L UN Combination Packaging, 4G/Class 6.2
Catalog No. 501534812
Encompass_Preferred
Biotransporter Cargo - 12L UN Combination Packaging, 4G/Class 6.2. Product is used with either code 437 or code 520 as the primary receptacles.
Catalog No. 50-153-4812 Supplier AirSea USA Supplier No. 424
Biotransporter Cargo - 12L UN Combination Packaging, 4G/Class 6.2. Product is used with either code 437 or code 520 as the primary receptacles.