Learn More
Bioworld bioTRAP Abundant Protein Removal Kit (Separate Antibodies), 4 mL
Supplier:  Bioworld 213400051
Encompass_Preferred
Abundant Protein Removalbio-WORLD's immunoaffinity columns will remove many of the abundant proteins in serum that interfere with proteomic analysis. These include albumin, transferrin, haptoglobin, alpha-1-antitrypsin, and many of the immunoglobins. By removing these proteins, the remaining serum can be concentrated, making detection of low abudance proteins easier. These columns are available packed with Separopore support. We also provide columns for the removal of any one of these proteins, or a custom mixture of depletion supports.Contains:1ml column with anti-albumin: Cat No. 22040002-11ml Column with anti-transferrin: Cat No. 22040003-11ml Column with anti-haptoglobin: Cat No. 22040001-11ml Column with alpha-1-trypsin: Cat No. 22040000-1Serum DiluentBinding/Washing Buffer Column Regeneration Solution
Catalog No. 50-198-594