Learn More
Bioworld BIOTRAP SERUM ABUNDANT PROT
Supplier:  Bioworld 213400002
Encompass_Preferred
BIOTRAP SERUM ABUNDANT PROTEIN REMOVAL KIT (MIXTURE OF ANTIBODIES): biotRAP Serum Abundant Protein Removal Kit bio-WORLD's biotRAP Serum Abundant Protein Removal kit removes four highly abundant proteins from human serum and plasma. Th...
Catalog No. 50-198-590