Learn More
Med Vet International BioVID 4K Microscope Camera, J0822BV4K, Each
Catalog No. 5000583975
Encompass_Preferred
Specifications: Resolution: 4K - 8 MegaPixels (3840 x 2160) Image Sensor: Sony IMX183 CMOS color 1/1.06"(13.06 x 7.27mm) Pixel size: 2.0um x 2.0um Auto-Exposure: 0.1ms-2000ms Power: cUL / CE approved power adapter AC Input: 100-240Vac 50/60Hz, 0.31A (Auto-switching power adapter) DC Output: 12V, 2A Adaptor Connections & Controls: USB Port: USB cable USB Mouse Port HDMI Port: HDMI cable to TV DC 12v Jack: 12v DC power in SD Slot: For included SD card ON/OFF: Power button LED Light: Red=off; Blue=on Requirements: Operating System: Microsoft Windows XP, Vista, 7, 8, 10 (32 & 64 bit), Mac OSX, Linux CPU: Intel Core2 2.8GHz or higher Memory: > 4GB USB High-speed port Weight and Dimensions: Height: 3.9" (98.3mm) Length: 2.6" (65mm) Width: 3.1" (78mm) Weight: 1.03lbs. (0.47kg) Video Output: HDMI User interface: USB mouse (included) Image/Video Capture: SD card Video: 30 frames per second @ 3840 x 2160 (HDMI) On screen-camera control panel and tool bar
Catalog No. 50-005-83975 Supplier Med Vet International Supplier No. JOR822BV4K
Specifications: Resolution: 4K - 8 MegaPixels (3840 x 2160) Image Sensor: Sony IMX183 CMOS color 1/1.06"(13.06 x 7.27mm) Pixel size: 2.0um x 2.0um Auto-Exposure: 0.1ms-2000ms Power: cUL / CE approved power adapter AC Input: 100-240Vac 50/60Hz, 0.31A (Auto-switching power adapter) DC Output: 12V, 2A Adaptor Connections & Controls: USB Port: USB cable USB Mouse Port HDMI Port: HDMI cable to TV DC 12v Jack: 12v DC power in SD Slot: For included SD card ON/OFF: Power button LED Light: Red=off; Blue=on Requirements: Operating System: Microsoft Windows XP, Vista, 7, 8, 10 (32 & 64 bit), Mac OSX, Linux CPU: Intel Core2 2.8GHz or higher Memory: > 4GB USB High-speed port Weight and Dimensions: Height: 3.9" (98.3mm) Length: 2.6" (65mm) Width: 3.1" (78mm) Weight: 1.03lbs. (0.47kg) Video Output: HDMI User interface: USB mouse (included) Image/Video Capture: SD card Video: 30 frames per second @ 3840 x 2160 (HDMI) On screen-camera control panel and tool bar