Learn More
BioSpec Products BioVortexer Kit
SDP
Catalog No. NC0400960
Encompass_Preferred
The BioVortexer™ is a battery-powered motor drive designed to rotate stirring rods or plastic pestles.  Rotating at thousands of revolutions per minute, it makes short work of dispersing centrifugal pellets or grinding small tissue samples. BioVortexer Kit, includes motor, batteries and 100 stirring rods.
Catalog No. NC0400960 Supplier BioSpec Products Supplier No. 1083MC
The BioVortexer™ is a battery-powered motor drive designed to rotate stirring rods or plastic pestles.  Rotating at thousands of revolutions per minute, it makes short work of dispersing centrifugal pellets or grinding small tissue samples. BioVortexer Kit, includes motor, batteries and 100 stirring rods.