Learn More
Research Products International Corp BioVortexer with 100 Stirring Rods for Micro-Centrifuge Tubes
SDP
Catalog No. 50212230
Encompass
The BioVortexer is a lightweight, battery powered unit designed to be used with microcentrifuge tubes. Centrifugal pellets, precipitates and other solids are completely dispersed in seconds by a combination of mechanical and vortex action. A new polypropylene stir rod is used with each sample.
A practical alternative to conventional vortex mixer for mixing and dispersing samples, the BioVortexer is faster, less tedious and safer. Take the BioVortexer to the sample instead of the sample to the mixer.
Disposable stirring rods are used to eliminate cross-contamination from sample to sample.
Catalog No. 50-212-230 Supplier Research Products International Corp Supplier No. 141290
The BioVortexer is a lightweight, battery powered unit designed to be used with microcentrifuge tubes. Centrifugal pellets, precipitates and other solids are completely dispersed in seconds by a combination of mechanical and vortex action. A new polypropylene stir rod is used with each sample.
A practical alternative to conventional vortex mixer for mixing and dispersing samples, the BioVortexer is faster, less tedious and safer. Take the BioVortexer to the sample instead of the sample to the mixer.
Disposable stirring rods are used to eliminate cross-contamination from sample to sample.