Learn More
MTC Bio BioWand™ Personal UV Sanitizer
Catalog No. NC1834174
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
MTCs new BioWand™ uses UV light (265 to 280nm) to disinfect any object/surface in the lab and beyond. BioWand™ can be set for a standard one-minute cleaning cycle or a more intense three-minute cycle. Simply press the power/start button and hold the light 1-2 inches from the surface and slowly move it over the entire target area. Allow the UV light to shine on the desired area for 5 to 10 seconds to ensure optimum exposure. The BioWand™ provides 2 hours of operating time and includes a USB-C cable for charging and portability.
Catalog No. NC1834174 Supplier MTC Bio Supplier No. U1001W
This item has a minimum qty of 3 per supplier requirements.
MTCs new BioWand™ uses UV light (265 to 280nm) to disinfect any object/surface in the lab and beyond. BioWand™ can be set for a standard one-minute cleaning cycle or a more intense three-minute cycle. Simply press the power/start button and hold the light 1-2 inches from the surface and slowly move it over the entire target area. Allow the UV light to shine on the desired area for 5 to 10 seconds to ensure optimum exposure. The BioWand™ provides 2 hours of operating time and includes a USB-C cable for charging and portability.