Learn More
Lonza™ BioWhittaker™ Dulbecco's Phosphate Buffered Saline (1X) w/o Calcium and Magnesium
Catalog No. BW17512Q12 Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 L
12 x 500 mL
24 x 500 mL
12 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW17512Q12 12 x 1 L
BW17-512Q 1 L
BW17512F24 24 x 500 mL
BW17512F12 12 x 500 mL
BW17-512F 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. BW17512Q12 Supplier Lonza™ Supplier No. 17512Q/12
Used to provide buffer system to maintain cell culture media in the physiological pH range of 7 to 7.6.
  • Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
  • Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dulbecco's Phosphate Buffered Saline
Without Additives No Calcium, No Magnesium
pH 7 to 7.6
Quantity 12 x 1 L