Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioWhittaker™ Water for Cell Culture, Lonza™

Catalog No. BW17724Q Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW17-724Q 1 L
BW17-724F 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BW17-724Q Supplier Lonza Inc Supplier No. 17724Q

Water for Injection (WFI) system for culture applications. BioWhittaker™ Water for Cell Culture is manufactured aseptically under cGMP conditions using a validated sterile filtration method.

  • Method for the production of WFI includes:
  • Ultra filtration
  • Reverse osmosis
  • Mixed bed deionizers
  • Hot recirculating loop and cooled at point of use
  • System is validated and monitored for bioburden, endotoxin, total organic carbons and conductivity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Sterility Sterile
Quantity 1 L
Chemical Name or Material Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.