Learn More
Medchemexpress LLC HY-18377 5mg Medchemexpress, Bioymifi CAS:1420071-30-2 Purity:>98%
SDP
Catalog No. 502082042
Encompass_Preferred
Medchemexpress, HY-18377 5mg Bioymifi CAS:1420071-30-2 Bioymifi (DR5 Activator), a potent TRAIL receptor DR5 activator, binds to the extracellular domain (ECD) of DR5 with a Kd of 1.2 μM. Bioymifi can act as a single agent to induce DR5 clustering and aggregation, leading to apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2042 Supplier Medchemexpress LLC Supplier No. HY183775MG
Medchemexpress, HY-18377 5mg Bioymifi CAS:1420071-30-2 Bioymifi (DR5 Activator), a potent TRAIL receptor DR5 activator, binds to the extracellular domain (ECD) of DR5 with a Kd of 1.2 μM. Bioymifi can act as a single agent to induce DR5 clustering and aggregation, leading to apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.