Learn More
Selleck Chemical LLC Bioymifi 5mg 1420071-30-2
SDP
Catalog No. 501365452
Encompass_Preferred
Bioymifi, a small-molecule death receptor 5 (DR5) agonist, binds to the extracellular domain(ECD) of DR5 with a Kd of 1.2 ?M but showed little binding affinity to the DR4 ECD. It induces DR5 clustering and aggregation, leading to apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5452 Supplier Selleck Chemical LLC Supplier No. S79425MG
Bioymifi, a small-molecule death receptor 5 (DR5) agonist, binds to the extracellular domain(ECD) of DR5 with a Kd of 1.2 ?M but showed little binding affinity to the DR4 ECD. It induces DR5 clustering and aggregation, leading to apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.