Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column50 x 2.1 mm

Catalog No. 502396394 Shop All Phenomenex Products
Encompass

bioZen™ 2.6 m Peptide XB-C18 00B-4768-AN

Catalog No. 50-239-6394 Supplier Phenomenex Inc Supplier No. 00B4768AN
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

bioZen™ 2.6 m Peptide XB-C18 00B-4768-AN

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.