Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Nano Column, 500 x 0.075 mm, Dual SecurityLink

Catalog No. 502396449 Shop All Phenomenex Products
Encompass

bioZen™ 5 m Peptide XB-C18 00J-4792-AW-22

Catalog No. 50-239-6449 Supplier Phenomenex Inc Supplier No. 00J4792AW22
May include imposed supplier surcharges.
Add to Cart
Add to Cart

bioZen™ 5 m Peptide XB-C18 00J-4792-AW-22

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.