Learn More
Bioworld bioZOL™ - M Microbial RNA Isolation, 100 mL
Supplier:  Bioworld 107600021
Encompass_Preferred
BioZOL-M Microbial RNA Isolation Kit is designed for the isolation of intact total RNA from Gram-positive or Gram-negative bacteria. It contains a bacterial wash reagent composed of chelating agents, detergent, Moreand a high ionic strength buffer, as a pre-treatment buffer for bacterial cells prior to RNA isolation. This reagent also inactivates RNase an denatures proteins to ensure good quality RNA.Haz Mat fee applies
Catalog No. 50-196-859