Learn More
Bioworld bioZOL™ - P, Plant RNA Reagent, 100 mL
Supplier:  Bioworld 107600031
Encompass_Preferred
bioZOL™-P, Plant RNA ReagentbioZOL™Kit is a single phase solution designed specifically for the isolation of pure, intact RNA from plant tissue and cells in less than 90 minutes Kit utilizes phenol, ammonium thiocyanate, guanidine thiocyanate, and B-mercaptoethanol to effectively disrupt cellular structure while inactivating RNase released from membrane-bound organelles Total RNA is free of proteinand DNA and can be used for cDNA synthesis, Northern blotting and in vitro translation Procedure eliminates the need for lengthy ethanol precipitations and cesium chloride gradients One kit performs 100 RNA isolations (each from 100 milligrams of tissue)Product can be used as a cost effective substitute to TRIzol
Catalog No. 50-196-860