Learn More
Medchemexpress LLC HY-111055 5mg Medchemexpress, BIP-135 CAS:941575-71-9 Purity:>98%
SDP
Catalog No. 501968817
Encompass_Preferred
Medchemexpress, HY-111055 5mg BIP-135 CAS:941575-71-9 BIP-135 is a potent and selective ATP-competitive GSK-3 inhibitor, with IC50s of 16 nM and 21 nM for GSK-3α and GSK-3β, respectively. BIP 135 exhibits neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8817 Supplier Medchemexpress LLC Supplier No. HY1110555MG
Medchemexpress, HY-111055 5mg BIP-135 CAS:941575-71-9 BIP-135 is a potent and selective ATP-competitive GSK-3 inhibitor, with IC50s of 16 nM and 21 nM for GSK-3α and GSK-3β, respectively. BIP 135 exhibits neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.