Learn More
Medchemexpress LLC BIP-135 | 941575-71-9 | 99.86% | 421.24 | 50 MG
SDP
Catalog No. 5000343434
Encompass_Preferred
BIP-135 is a potent and selective ATP-competitive GSK-3 inhibitor, with IC50s of 16 nM and 21 nM for GSK-3α and GSK-3β, respectively. It exhibits a neuroprotective effect.
  • Potent and selective ATP-competitive GSK-3 inhibitor
  • IC50 for GSK-3α: 16 nM
  • IC50 for GSK-3β: 21 nM
  • Increases survival motor neuron (SMN) protein levels in human SMA fibroblasts
  • Neuroprotective agent in models of oxidative stress
  • Well-tolerated in animal studies; non-toxic
  • Prolongs median survival time in SMA mouse models
Catalog No. 50-003-43434 Supplier Medchemexpress LLC Supplier No. HY11105550MG
BIP-135 is a potent and selective ATP-competitive GSK-3 inhibitor, with IC50s of 16 nM and 21 nM for GSK-3α and GSK-3β, respectively. It exhibits a neuroprotective effect.
  • Potent and selective ATP-competitive GSK-3 inhibitor
  • IC50 for GSK-3α: 16 nM
  • IC50 for GSK-3β: 21 nM
  • Increases survival motor neuron (SMN) protein levels in human SMA fibroblasts
  • Neuroprotective agent in models of oxidative stress
  • Well-tolerated in animal studies; non-toxic
  • Prolongs median survival time in SMA mouse models